Tlačiť
Reagent reservoir, Dual Solution®
Dodávateľ: HEATHROW
HS20821A
HEATHS20821AEA 122.17 EUR
HEATHS20821A
Reagent reservoir, Dual Solution®
Zásobníky na reagencie
PP. This non sterile solution basin/reagent reservoir has a standard 50 ml basin on one side and 12 individual 5 ml basins on the other side.
  • Wells are angled both downward and outward guiding mult ichannel pipette tips toward a centered position
  • Avoids contamination and waste
  • Imprinted indices identify samples
  • Loose-fitting lids prevent evaporation and contamination