Tlačiť
Multifunctional cooling thermoshakers, MKR 13 and MKR 23
Dodávateľ: HAEP LABOR
980123003
HAEP980123003EA 4307.63 EUR
HAEP980123003
Multifunctional cooling thermoshakers, MKR 13 and MKR 23
Třepačky Třepačky inkubované
Cooling thermomixers featuring the traditional orbital shaking motion and active counter-cooling. Holds one or two blocks. Versatile, suitable for a huge range of applications. For test tubes, sample jars, centrifuge tubes, cuvettes or PCR plates, microtitre and deep well plates. High-performance, hard-wearing Peltier elements guarantee the fastest possible tempering times and a long service life. Effective cooling with speed-controlled ventilators discharge excess heat or cold. Features a timer, short-mix function and interval shaking with shock effect.
  • Housing made of hard anodised
More Product Information