Tlačiť
Reference electrode for use with ion selective electrodes, sensION+ 5044
Dodávateľ: HACH
LZW5044970002
HACHLZW5044970002EA 472.66 EUR
HACHLZW5044970002
Reference electrode for use with ion selective electrodes, sensION+ 5044
Elektrody Elektrody ISE
The 5044 with lithium acetate electrolyte is recommended for use with all sensION+ half-cell ion selective electrodes (ISE) in drinking water, wastewater and general water quality applications.
  • Double junction reference electrode with internal gel electrolyte and 0,1 M lithium acetate salt bridge
  • Guarded against contamination
  • Quick measurement stabilization with large ceramic junction
  • Easy to maintain: Lithium acetate electrolyte is refillable
  • Fixed 1 meter cable and banana connector for laboratory use with the Hach sensION+ MM374 Multimeter