Tlačiť
Test kits, photometric, Chromium, LCK cuvette tests
Dodávateľ: Hach
LCK313
HACHLCK313EA 186.26 EUR
HACHLCK313
Test kits, photometric, Chromium, LCK cuvette tests
Kity pro environmentální testy
Pre-dosed reagents for maximum safety.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 2 to 8 °C (keep refrigerated)
Certifikácie: EN ISO 11083, DIN 38405-D24