Tlačiť
Ion selective electrode, Ammonia, solid-state, IntelliCAL™
Dodávateľ: HACH
ISENH418101 ISENH418103 ISENH4181AP
HACHISENH418101EA 1360.3 EUR
HACHISENH418101 HACHISENH418103 HACHISENH4181AP
Ion selective electrode, Ammonia, solid-state, IntelliCAL™
Elektrody Elektrody ISE
IntelliCAL™ ISENH4181 is a digital combination ammonium ion selective electrode (ISE) with a non refillable double junction reference and built-in temperature sensor. The solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Provides fast, stable, and accurate response
  • Ultimate traceability in measurement history
  • User alerted when re-calibration is needed
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings