Tlačiť

EDTA 0.1428 mol/l aqueous solution

Dodávateľ: Hach

Synonymá: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II

1496001
HACH1496001EA 47.27 EUR
HACH1496001
EDTA 0.1428 mol/l aqueous solution
EDTA
Vzorec: C₁₀H₁₆N₂O₈
Mw: 292.25
Teplota varu: 614
Teplota tavenia: 220
Skladovacia teplota: Ambient
Číslo MDL: MFCD00003541
Číslo CAS: 60-00-4
EINECS: 200-449-4
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR