Tlačiť
Heat exchanger and circulator packages
Dodávateľ: HAAKE
222-1937
HAAK222-1937EA 6479.87 EUR
HAAK222-1937
Heat exchanger and circulator packages
Termostaty oběhové Termostaty oběhové Termostat cirkulační s ohřevem
SC100- A10 refrigerated bath circulator provides efficient cooling for open and closed applications, with temperatures ranging from −10 to +100°C and maximum bath volume of 6 litres.
  • Powerful bath circulator with a stainless steel reservoir
Informace o dodávce: Circulator package includes SC100-A10 and 3 m silicon hose