Tlačiť
Transport boxes, VACUETTE®
Dodávateľ: Greiner Bio-One
472001_10
GREI472001_10EA 268.78 EUR
GREI472001_10
Transport boxes, VACUETTE®
Krabice Krabice přepravní
Transport box made of PP.
  • For transport of up to 40 tubes of diameter 9 to 16 mm
  • With practical snap closure
  • Leak-proof, due to sealing ring and absorbent pad
  • Stackable
  • Separate slot for cooler pack
Ensure the safe transport of sample material from the collection point right up to analysis in the laboratory.