Tlačiť
Microplate sealing tape
Dodávateľ: G BIOSCIENCES
786-422
GENO786-422EA 418.59 EUR
GENO786-422
Microplate sealing tape
Těsnění Těsnění mikrodestičky
Acetate film with acrylic adhesive. This self-adhesive microplate sealing tape is suitable for sealing both 96-well and 384-well plates. The precut, pressure-senstive tape seals and reseals the surface of the plates during incubations to prevent evaporation and allow more vigorous mixing. The tape will reseal the plates 2 to 3 times.
  • Non sterile
  • Transparent and non pierceable
  • Minimise evaporation and condensation
  • For short and long term storage of plates
  • Temperature range: −16 to +38 °C