Tlačiť
Normal Human Serum
Dodávateľ: GEMINI BIO-PRODUCTS
100-110-100
GEMB100-110-100EA 255.96 EUR
GEMB100-110-100
Normal Human Serum
Séra
Normal human serum is collected from healthy male and female donors of all ABO serotypes at FDA-licensed facilities in the United States. This material is defibrinated from source plasma. All donor units are tested for viral markers and found to be non-reactive.
  • 0,1 µm sterile-filtered
Informace o dodávce: Ships on dry ice.