Tlačiť
Permanent Autoclave-Resistant Thermal Transfer Labels
Dodávateľ: GA
AUTC-258C1-3BL AUTC-258C1-3GA AUTC-258C1-3RE AUTC-258C3-6BL AUTC-258C3-6GA AUTC-258C3-6RE AUTC75NPC1-2GA AUTC75NPC1-2OR AUTC75NPC3-2GA AUTC75NPC3-2OR AUTC84C10.25BL AUTC84C10.25GA AUTC84C10.25OR AUTC84C10.25PI AUTC84C10.25RE AUTC84C10.25YE AUTC-84C3-1BL AUTC-84C3-1GA AUTC-84C3-1OR AUTC-84C3-1PI AUTC-84C3-1RE AUTC-84C3-1YE AUTT-103C1-2WH AUTT-103C3-2WH AUTT-105C1-2WH AUTT-105C3-2WH AUTT-19C1-1WH AUTT-19C3-1WH AUTT-200C1-1WH AUTT-200C3-2WH AUTT216C10.5WH AUTT-216C3-1WH AUTT-231C3-2WH AUTT-258C1-3WH AUTT-258C3-6WH AUTT-27C1-1WH AUTT-27C3-2WH AUTT-28C1-2WH AUTT-28C3-2WH AUTT-36C1-1WH AUTT62C1-0.5WH AUTT-62C3-1WH AUTT75NPC1-2WH AUTT75NPC3-2WH AUTT84C10.25WH AUTT-84C3-1WH
GAINAUTC-258C1-3BLEA 191.87 EUR
GAINAUTC-258C1-3BL GAINAUTC-258C1-3GA GAINAUTC-258C1-3RE GAINAUTC-258C3-6BL GAINAUTC-258C3-6GA GAINAUTC-258C3-6RE GAINAUTC75NPC1-2GA GAINAUTC75NPC1-2OR GAINAUTC75NPC3-2GA GAINAUTC75NPC3-2OR GAINAUTC84C10.25BL GAINAUTC84C10.25GA GAINAUTC84C10.25OR GAINAUTC84C10.25PI GAINAUTC84C10.25RE GAINAUTC84C10.25YE GAINAUTC-84C3-1BL GAINAUTC-84C3-1GA GAINAUTC-84C3-1OR GAINAUTC-84C3-1PI GAINAUTC-84C3-1RE GAINAUTC-84C3-1YE GAINAUTT-103C1-2WH GAINAUTT-103C3-2WH GAINAUTT-105C1-2WH GAINAUTT-105C3-2WH GAINAUTT-19C1-1WH GAINAUTT-19C3-1WH GAINAUTT-200C1-1WH GAINAUTT-200C3-2WH GAINAUTT216C10.5WH GAINAUTT-216C3-1WH GAINAUTT-231C3-2WH GAINAUTT-258C1-3WH GAINAUTT-258C3-6WH GAINAUTT-27C1-1WH GAINAUTT-27C3-2WH GAINAUTT-28C1-2WH GAINAUTT-28C3-2WH GAINAUTT-36C1-1WH GAINAUTT62C1-0.5WH GAINAUTT-62C3-1WH GAINAUTT75NPC1-2WH GAINAUTT75NPC3-2WH GAINAUTT84C10.25WH GAINAUTT-84C3-1WH
Permanent Autoclave-Resistant Thermal Transfer Labels
Štítky
Permanent autoclave-resistant labels withstand various sterilisation procedures, including in steam autoclaves, dry heat ovens, gamma irradiators, and ethylene oxide gas sterilisers.
  • Temperature range: −60 to 150 °C
  • Ideal for use in repeated steam autoclave sterilization cycles, gamma irradiation (up to 50 KGy), and ethylene oxide sterilisation
  • Printable with all major thermal transfer printer brands
  • Waterproof and weatherproof
  • RR-class resin ribbon recommended
High temperature-resistant permanent labels for identifying various sterilisationMore Product Information