Tlačiť
Laser Particle Sizer, ANALYSETTE 22 NeXT Nano
Dodávateľ: FRITSCH
22.9040.00
FRIT22.9040.00EA 0 EUR
FRIT22.9040.00
Laser Particle Sizer, ANALYSETTE 22 NeXT Nano
Analyzátory Analyzátory prvků
ANALYSETTE 22 NeXT Nano comes with an extra wide measuring range of 0,01 to 3800 μm for maximum precision and sensitivity for smallest particles with an additional detector system.
  • Easy operation and cleaning
  • Short analysis times
  • Consistent reproducibility, reliable comparability
  • Solid and low-maintenance with few moving parts
  • Only one light source: simple, robust, reliable
  • Quick and simultaneous recording of all scattering data
  • Highly precise recording of the light intensities by state-of-the-art 16 bit converter technology
  • Recording also of very wide
More Product Information