Tlačiť
Restriction enzymes, DraI, FastDigest™, Fermentas
Dodávateľ: Thermo Fisher Scientific
FD0224
FD0224EA 96.94 EUR
FD0224
Restriction enzymes, DraI, FastDigest™, Fermentas
Činidla pro nukleové kyseliny Restriction Enzymes
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications
Designed to complete digestion in 5 to 15 minutes.