Tlačiť
Avantor® ACE® Excel® CN-ES, HPLC/UHPLC Columns, Analytical, 5 µm
Dodávateľ: Avantor
EXL-1213-1002UEA 855.2 EUR
EXL-1213-1002U EXL-1213-1003U EXL-1213-1046U EXL-1213-1502U EXL-1213-1503U EXL-1213-1546U EXL-1213-2502U EXL-1213-2503U EXL-1213-2546U EXL-1213-0502U EXL-1213-0503U EXL-1213-0546U
Avantor® ACE® Excel® CN-ES, HPLC/UHPLC Columns, Analytical, 5 µm
Kolony chromatografické
Avantor® ACE® CN-ES combines CN polar selectivity with enhanced hydrophobicity, enabling the benefits of both interactions to be fully exploited. Extra phase stability and increased column lifetime is provided by an extended alkyl chain spacer between the silica surface and cyano group. Avantor® ACE® CN-ES is suitable for both reversed phase and normal phase separations.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • With hydrophobic plus CN polar selectivity, especially useful for polar molecules
  • Wide range of particle sizes (1
More Product Information