Tlačiť
Avantor® ACE® Excel® C8, HPLC/UHPLC Columns, Analytical, 2 µm
Dodávateľ: Avantor
EXL-102-1002UEA 950.93 EUR
EXL-102-1002U EXL-102-1003U EXL-102-1046U EXL-102-1203U EXL-102-1502U EXL-102-1503U EXL-102-1546U EXL-102-0302U EXL-102-0502U EXL-102-0503U EXL-102-0546U EXL-102-7502U EXL-102-7503U EXL-102-7546U ACE-1P2-GD2U ACE-1P2-GD4U
Avantor® ACE® Excel® C8, HPLC/UHPLC Columns, Analytical, 2 µm
Kolony chromatografické
These ultra-inert Avantor® ACE® Excel® C8 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 5 µm) and complementary phases
  • Can be used as general purpose starting point column
  • Method validation kits contain three different batches of the same phase -
More Product Information