Tlačiť
TDS testers, Eutech EcoTestr TDS High / TDS Low
Dodávateľ: EUTECH
ECOTDSTESTLOW ECOTDSTESTHIGH
EUTEECOTDSTESTLOWEA 104.55 EUR
EUTEECOTDSTESTLOW EUTEECOTDSTESTHIGH
TDS testers, Eutech EcoTestr TDS High / TDS Low
Meřiče elektrochemické Konduktometry
These waterproof testers come with adjustable TDS factor, allowing users to measure TDS in a wide range of applications. EcoTestr TDS High measures from 0 to 10 ppt, EcoTestr TDS Low measures from 0 to 1990 ppm.
  • Adjustable TDS factor from 0,4 (Low) or 0,5 (High) to 1,0
  • Hold function freezes reading temporarily for ease of recording
  • IP67 waterproof – tester floats on water for easy retrieval
  • Long sensor life – no need to change sensors
  • Transparent protective cap that doubles up as a container for samples or calibration solution
  • Large, upright LCD with
More Product Information