Tlačiť
Carrying kit for meter and electrodes, Eutech
Dodávateľ: EUTECH
ECECOPHKIT
EUTEECECOPHKITEA 89.73 EUR
EUTEECECOPHKIT
Carrying kit for meter and electrodes, Eutech
Meřiče elektrochemické Meřiče pH a multimetry
Made of sturdy reinforced plastic, the kit comes complete with calibrating sachets and sample bottles, and is thoughtfully designed to hold one meter and electrodes.