Tlačiť
Insulin ELISA kit
Dodávateľ: ENZO LIFE SCIENCES
ENZKIT1410001
ENZOENZKIT1410001EA 588.02 EUR
ENZOENZKIT1410001
Insulin ELISA kit
Test ELISY
Highly sensitive Insulin ELISA kit enabling detection of insulin in serum, plasma, tissue culture media in just 3 hours.
  • Highly sensitive measurement of insulin, detecting as little as 21,6 pg/ml (0,622 µIU/ml)
  • Negligible cross reactivity with proinsulin
  • High throughput format with results in 3 hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of insulin in serum, plasma, and tissue culture media. The Insulin ELISA kit is a colorimetric immunometric immunoassay kit with results in 3 hours.
Objednávacie informácie: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.