Tlačiť
Z-Leu-Leu-Glu-Amc
Dodávateľ: ENZO LIFE SCIENCES
BMLZW93450005
ENZOBMLZW93450005EA 195.47 EUR
ENZOBMLZW93450005
Z-Leu-Leu-Glu-Amc
Proteiny
Fluorogenic substrate for the peptidylglutamyl-peptide hydrolysing (caspase-like) activity of the proteasome which may be stimulated in the presence of Mg2+ ions and rapidly inactivated by N-acetylimidazole.