Tlačiť
PDKtide, biotin
Dodávateľ: ENZO LIFE SCIENCES
BMLP2510100
ENZOBMLP2510100EA 259.16 EUR
ENZOBMLP2510100
PDKtide, biotin
Proteiny
Protein kinase substrate
This peptide, Biotin-Ahx-KTFCGTPEYLAPEVRREPRILSEEEQEMFRDFDYIADWC, is derived from AKT1 and PKN2/PRK2. It is a substrate for PDK1, CDK9/Cyclin T1 and JAK2.The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes.