Tlačiť

Gambogic amide 98 TLC_ASSAY_METHOD

Dodávateľ: ENZO LIFE SCIENCES
Nebezpečí

BMLN1590001 BMLN1590005
ENZOBMLN1590001EA 188.66 EUR
ENZOBMLN1590001 ENZOBMLN1590005
Gambogic amide 98 TLC_ASSAY_METHOD
Gambogic amide

TrkA ligand


A novel small molecule NGF mimic discovered using a cell-based chemical genetic screen for TrkA agonists. Gambogic amide selectively binds to TrkA, triggers its tyrosine phosphorylation, elicits PI3-kinase/Akt and MAPK activation, provokes neurite outgrowth in PC12 cells and prevents neuronal cell death. EC50 for antiapoptotic activity is 5nM.


Soluble in DMSO (20 mg/ml) or ethanol (20 mg/ml).

Vzorec: C₃₈H₄₅NO₇
Mw: 627.8 g/mol
Teplota tavenia: 74…76 °C
Skladovacia teplota: Ambient
Číslo CAS: 286935-60-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ede2973f64c24278e3fee48bc4f9bcbecc0b0af7&vid=0bf45e22b3a568f469bd8d5855fb9568db7bdbd8&prid=web&clid=SAPDAM

Order Now

Výsledky špecifikačného testu

Purity ≥98% (TLC)
Appearance Yellow-orange solid
Melting Point 74 - 76°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR