Tlačiť

(+/-)-Anatoxin ≥98% (by TLC)

Dodávateľ: ENZO LIFE SCIENCES
Nebezpečí

BMLC1180001
ENZOBMLC1180001EA 349.69 EUR
ENZOBMLC1180001
(+/-)-Anatoxin ≥98% (by TLC)
(+/-)-Anatoxin

Nicotinic acetylcholine receptor ligand.


(±)2-Acetyl-Anatoxin is an alkaloidal toxin produced by the filamentous freshwater cyanophyte Anabaena flos-aquae. This compound is the synthetic version.


(±)-Anatoxin is a potent agonist at neuronal nicotinic acetylcholine receptors. At low concentrations (20 to 200 nM) it acts as an agonist. At higher concentrations (>800 nM), it blocks the AChR ion channel, desensitises the receptor and blocks neuromuscular transmission.


Soluble in water (14 mg/ml).


Pozor: Research use only. Store, as supplied, at –20 °C for up to 1 year. Store solutions at –20 °C for up to 1 month.

Vzorec: C₁₀H₁₅NO·C₄H₄O₄
Mw: 281.3 g/mol
Skladovacia teplota: Freezer
Číslo CAS: 1219922-30-1
OSN: 3462
ADR: 6.1 I

Order Now

Výsledky špecifikačného testu

Purity ≥98% (TLC)
Appearance White solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR