Tlačiť

Anti-CD42C Mouse Monoclonal Antibody

Dodávateľ: ENZO LIFE SCIENCES
ALX805026C100
ENZOALX805026C100EA 449.83 EUR
ENZOALX805026C100
Anti-CD42C Mouse Monoclonal Antibody
Protilátky
Host: Mouse, Isotype: IgG2b

  • For Immunoprecipitation, Western Blot

Recognizes the human GPIbbeta subunit (CD42c) of the human GPIb/IX/V (CD42a/CD42b/CD42c/CD42d) complex.

Type: Primary
Antigen: CD42C
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
New Product
Order Now

ŠPECIFIKÁCIA

Symbol antigenu CD42C
Environmentally Preferable
Druh protilátky Primární
Klonovalita Monoclonal
Reaktivita
Hostitel Mouse
Izotyp IgG2b
Western blot Yes
Imunoprecipitace Yes
Formulář Liquid. 100 µg of purified antibody in 200 µl PBS containing 0,05% sodium azide
Čištění 0.95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR