Tlačiť

Anti-MDA5 Mouse Monoclonal Antibody [clone: 1B6]

Dodávateľ: ENZO LIFE SCIENCES
ALX804878C100
ENZOALX804878C100EA 456.64 EUR
ENZOALX804878C100
Anti-MDA5 Mouse Monoclonal Antibody [clone: 1B6]
Protilátky
MDA5 is a highly conserved helicase whose ATPase activity can be stimulated by RNA and less potently by DNA. It contains two N-terminal CARD domains and a C-terminal helicase domain. The helicase domain, once freed from the CARD domains, then translocates from the cytoplasm to the nucleus where it acts on chromatin architecture. This could allow easier access of CAD, thereby accelerating the degradation of genomic DNA.

Type: Primary
Antigen: MDA5
Clonality: Monoclonal
Clone: 1B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR