Tlačiť
omega-Agatoxin IVA
Dodávateľ: ENZO LIFE SCIENCES
ALX630001C100
ENZOALX630001C100EA 911.28 EUR
ENZOALX630001C100
omega-Agatoxin IVA
Proteiny
Ca²⁺ channel blocker
Selective P-type Ca²⁺ channel blocker. Fast acting and potent neurotoxin.