Tlačiť
Neutralising reagent
Dodávateľ: ENZO LIFE SCIENCES
ADI801475
ENZOADI801475EA 84.52 EUR
ENZOADI801475
Neutralising reagent
Pufry Pufry biologické
Neutralising buffer for low pH solutions and for use with cyclic nucleotide and isoprostane kits.