Tlačiť
Inline gas filters, Wafergard® II F Micro
Dodávateľ: ENTEGRIS
WG2FT1RF2 WG2FT1RR2 WG2FT1SS2 WGFG01RH2 WG2FT1BW2
ENTEWG2FT1RF2EA 0 EUR
ENTEWG2FT1RF2 ENTEWG2FT1RR2 ENTEWG2FT1SS2 ENTEWGFG01RH2 ENTEWG2FT1BW2
Inline gas filters, Wafergard® II F Micro
Filtry Filtry na plyny
Recommended for inert and reactive gases, these filters offer excellent compatibility with all classes of semiconductor process gases. Its compact diameter enables easy installation in cramped gas panels and cabinets.
  • Superior particulate filtration for ultrapure gas system filtration
  • Low pressure drop reduces the risk of condensation in low vapour pressure gases
  • Cleanroom manufactured, welded and tested
  • PTFE membranes provide high efficiency filtration
Filter element: hydrophobic PTFE membrane supported by moulded PTFE PFA structure; housing:More Product Information