Tlačiť
Cartridge filters, Fluorogard® FP
Dodávateľ: ENTEGRIS
CWEG010E1 CWEG010V1 CWEV010T1 CWEG010T1 CWEA010E1 CWEA010T1 CWEA010V1 CWEA020E1 CWEA020T1 CWEA020V1 CWEA030T1 CWEA710E1 CWEC010E1 CWEC010T1 CWEC020T1 CWEC710E1 CWEG010T1Y CWEG020E1 CWEG020T1 CWEG020V1 CWEG030E1 CWEG030T1 CWEG710E1 CWEG710V1 CWEG720E1 CWEH010E1 CWEH010T1 CWEH010T1Y CWEH020T1 CWEH030E1 CWEH030T1 CWES010E1 CWES010T1 CWES030E1 CWES710E1 CWES710T1 CWES720E1 CWEV010E1 CWEV010T1Y CWEV010V1 CWEV020E1 CWEV020T1 CWEV030T1 CWEV710E1 CWEV720E1 CWEZ010E1 CWEZ010T1 CWEZ020T1 CWEAF10E1 CWEAF20E1 CWEAF30E1 CWECF10E1 CWEGF10E1 CWEGF20E1 CWEHF10E1 CWESF10E1 CWESF20E1 CWEVF10E1 CWEZF10E1
ENTECWEG010E1EA 0 EUR
ENTECWEG010E1 ENTECWEG010V1 ENTECWEV010T1 ENTECWEG010T1 ENTECWEA010E1 ENTECWEA010T1 ENTECWEA010V1 ENTECWEA020E1 ENTECWEA020T1 ENTECWEA020V1 ENTECWEA030T1 ENTECWEA710E1 ENTECWEC010E1 ENTECWEC010T1 ENTECWEC020T1 ENTECWEC710E1 ENTECWEG010T1Y ENTECWEG020E1 ENTECWEG020T1 ENTECWEG020V1 ENTECWEG030E1 ENTECWEG030T1 ENTECWEG710E1 ENTECWEG710V1 ENTECWEG720E1 ENTECWEH010E1 ENTECWEH010T1 ENTECWEH010T1Y ENTECWEH020T1 ENTECWEH030E1 ENTECWEH030T1 ENTECWES010E1 ENTECWES010T1 ENTECWES030E1 ENTECWES710E1 ENTECWES710T1 ENTECWES720E1 ENTECWEV010E1 ENTECWEV010T1Y ENTECWEV010V1 ENTECWEV020E1 ENTECWEV020T1 ENTECWEV030T1 ENTECWEV710E1 ENTECWEV720E1 ENTECWEZ010E1 ENTECWEZ010T1 ENTECWEZ020T1 ENTECWEAF10E1 ENTECWEAF20E1 ENTECWEAF30E1 ENTECWECF10E1 ENTECWEGF10E1 ENTECWEGF20E1 ENTECWEHF10E1 ENTECWESF10E1 ENTECWESF20E1 ENTECWEVF10E1 ENTECWEZF10E1
Cartridge filters, Fluorogard® FP
Filtry Patrony filtrační
These filters can be used in solvent based photochemicals without the need for prewetting steps.
  • Excellent chemical compatibility
  • For use in POU filtration and chemical delivery system filtration
Membrane: Hydrophobic PTFE; Supports, core, sleeve and endcaps: PP
Maximum operating temperature: 90 °C
Maximum forward/reverse differential pressure: 3,4 bar at 25 °C