Tlačiť
Test sieves, 200×50 mm
Dodávateľ: ENDECOTTS
200SAW.020 200SAW.025 200SAW.032 200SAW.038 200SAW.045 200SAW.053 200SAW.063 200SAW.075 200SAW.090 200SAW.106 200SAW.125 200SAW.150 200SAW.180 200SAW.212 200SAW.250 200SAW.300 200SAW.355 200SAW.425 200SAW.500 200SAW.600 200SAW.710 200SAW.850 200SAW1.00 200SAW1.18 200SAW1.40 200SAW1.70 200SAW100 200SAW106 200SAW11.2 200SAW12.5 200SAW125 200SAW13.2 200SAW16.0 200SAW19.0 200SAW2.00 200SAW2.36 200SAW2.80 200SAW22.4 200SAW25.0 200SAW26.5 200SAW3.35 200SAW31.5 200SAW37.5 200SAW4.00 200SAW4.75 200SAW45.0 200SAW5.60 200SAW50.0 200SAW53.0 200SAW6.30 200SAW6.70 200SAW63.0 200SAW75.0 200SAW8.00 200SAW9.50 200SAW90.0
ENDE200SAW.020EA 1229.72 EUR
ENDE200SAW.020 ENDE200SAW.025 ENDE200SAW.032 510-0356 510-0357 ENDE200SAW.053 510-0358 510-0359 510-0360 ENDE200SAW.106 510-0361 ENDE200SAW.150 510-0362 ENDE200SAW.212 510-0363 510-0364 510-0365 510-0366 510-0367 ENDE200SAW.600 510-0368 510-0369 510-0370 ENDE200SAW1.18 ENDE200SAW1.40 ENDE200SAW1.70 ENDE200SAW100 ENDE200SAW106 ENDE200SAW11.2 ENDE200SAW12.5 ENDE200SAW125 ENDE200SAW13.2 ENDE200SAW16.0 ENDE200SAW19.0 510-0371 ENDE200SAW2.36 ENDE200SAW2.80 ENDE200SAW22.4 ENDE200SAW25.0 ENDE200SAW26.5 ENDE200SAW3.35 ENDE200SAW31.5 ENDE200SAW37.5 ENDE200SAW4.00 ENDE200SAW4.75 ENDE200SAW45.0 ENDE200SAW5.60 ENDE200SAW50.0 ENDE200SAW53.0 ENDE200SAW6.30 ENDE200SAW6.70 ENDE200SAW63.0 ENDE200SAW75.0 ENDE200SAW8.00 ENDE200SAW9.50 ENDE200SAW90.0
Test sieves, 200×50 mm
Síta Síta
These test sieves are made from stainless steel.
Certifikácie: According to ASTM E 11-95
Informace o dodávce: Each sieve is delivered with an individual quality certificate.