Tlačiť
ESD Safety shoe WHITE GRIP Low ESD S2, Upper material: Hydrophobized microfibre, Full-length inlay sole SPORTIVE ESD, Soft fleece ES D insole, sole: RUBBER/PU E-TRACK, Toe cap: steel, color: white, Size EU39 / UK6 1 * 1 PAIR
Dodávateľ: ELTEN