Tlačiť

Tris(perfluorobutyl)amine

Dodávateľ: EHRENSTORFER

Synonymá: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine

C15987500
EHERC15987500EA 97.74 EUR
EHERC15987500
Tris(perfluorobutyl)amine
Tris(perfluorobutyl)amine
Vzorec: C₁₂F₂₇N
Mw: 671.1
Teplota varu: 155
Teplota tavenia: –55 °C
Hustota: 1.883 g/cm³ (20 °C)
Číslo MDL: MFCD00000436
Číslo CAS: 311-89-7
EINECS: 206-223-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR