Tlačiť
Kits: cholesterol diagnostics, Ready-to-Load™
Dodávateľ: EDVOTEK
118 118-C
EDVO118EA 113.36 EUR
EDVO118 EDVO118-C
Kits: cholesterol diagnostics, Ready-to-Load™
Materiály výukové Materiál pro výuku biologie Aktivity pro výuku genetiky
Genetic testing can be used to identify people with a genetic condition which caused them to have an elevated level of cholesterol and which can be fatal. Students can see how genetic testing is carried out and learn about DNA electrophoresis.
  • For six lab groups
Also required: Electrophoresis tank, power supply, micropipettes (5 to 50 µl adjustable or 40 µl fixed volume) with tips, and a white light box is recommended.
Informace o dodávce: Kit includes instructions, Ready-to-Load™ DNA samples, agarose powder, practice gel loading solution,More Product Information