Tlačiť
BlueLine Insulated Bags
Dodávateľ: DELTA T
B100210 B100230 B100205 B100203 B100216
DLTTB100210EA 95.73 EUR
DLTTB100210 DLTTB100230 DLTTB100205 DLTTB100203 DLTTB100216
BlueLine Insulated Bags
Sáčky Sáčky skladovací
BlueLine Bags are economical and durable. Its insulating property ensures the maintainance of constant internal temperature and protects the samples against heat or cold environment.
  • Bags has wide application in clinical studies, blood transport, medications, pharmaceutical and biotechnology sectors
  • TempShells and TempFrames elements fit in the bag perfectly without wastage of space
BlueLine bag 3 L: Suitable for 1 pair TempShell element and 0,5 pair of TempShell frame, with document pocket
BlueLine bag 5 L: Suitable for 2 pair of TempShell elements orMore Product Information