Tlačiť
Alkaline Batteries
Dodávateľ: DIALSTAT OFFICE SUPPLIES
EA6752
DIALEA6752EA 109.35 EUR
DIALEA6752
Alkaline Batteries
Baterie
Duracell Industrial batteries are designed for industrial-strength power. These AA batteries are ideal for use in torches, safety equipment, communications devices, telemeters, medical instruments and more. They are designed for extreme usage.
  • Capable of operation in temperatures from –20 to 54 °C
  • Storage up to seven years