Tlačiť
Hemoglobin standard
Dodávateľ: Cayman Chemical
700543-1
CAYM700543-1EA 28.24 EUR
CAYM700543-1
Hemoglobin standard
Standardy Standardy biologické
Ready for use with colorimetric assay kit. This vial contains solution of 500 µM heme.
Stability: ≥1 year at 4 °C.
Pozor: For laboratory use only. Not for human or veterinary use.