Tlačiť

Riboflavin 5'-monophosphate sodium salt hydrate

Dodávateľ: Cayman Chemical

18167-10 18167-100 18167-25 18167-50
CAYM18167-10EA 54.08 EUR
CAYM18167-10 CAYM18167-100 CAYM18167-25 CAYM18167-50
Riboflavin 5'-monophosphate sodium salt hydrate
Enzymy

Riboflavin 5'-monophosphate (FMN) is a coenzyme that is tightly bound to enzymes catalysing oxidation and reduction reactions in a variety of biosynthetic pathways.


  • Soluble in organic solvents - ethanol; DMSO and DMF
  • Solubility in PBS is 10 mg/ml


FMN also binds the FMN riboswitch (RFN element) on RNA to alter gene regulation. FMN is a substrate of FMN phosphohydrolases and is used to study their function.


Synonyms: Riboflavin 5'-(dihydrogen phosphate), monosodium salt, dihydrate; flavin mononucleotide, FMN, vitamin B2 phosphate.

Order Now

ŠPECIFIKÁCIA

Název enzymu Riboflavin 5'-monophosphate sodium salt hydrate
Environmentally Preferable
Zdroj
Druhy
Molekulární hmotnost 514,4 g/mol
Čistota ≥95%
Podmínky uskladnění −20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR