Tlačiť

Anti-CD3-delta Rabbit Polyclonal Antibody [clone: T56]

Dodávateľ: BIOWORLD TECHNOLOGY
BS3548
BWRLBS3548EA 321.65 EUR
BWRLBS3548
Anti-CD3-delta Rabbit Polyclonal Antibody [clone: T56]
Protilátky
Synthetic peptide, corresponding to amino acids 21-70 of Human CD3-delta.

CD3-? (T56) pAb detects endogenous levels of CD3-? protein.

Type: Primary
Antigen: CD3-delta
Clonality: Polyclonal
Clone: T56
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

ŠPECIFIKÁCIA

Symbol antigenu CD3- delta
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Klon T56
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Izotyp
Western blot Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR