Tlačiť
Immersion cylinder
Dodávateľ: BURKLE
5365-6000
BURK5365-6000EA 619.27 EUR
BURK5365-6000
Immersion cylinder
Vzorkovače Vzorkovače kapalné
Precise liquid samples can be taken with this immersion cylinder. When the target has been reached, open the cover and pour out the sample.
  • Screw-off top enables easy cleaning
  • Air valve at the end of the tube
  • Venting lowering cable and cable reel