Tlačiť
Amyloid plaque stain reagent, with EtBr counter stain, Amylo-Glo® RTD™
Dodávateľ: Biosensis
TR-400-AG
BSENTR-400-AGEA 527.14 EUR
BSENTR-400-AG
Amyloid plaque stain reagent, with EtBr counter stain, Amylo-Glo® RTD™
Test Buněčná stanovení
Amylo-Glo RTD reagent is designed to stain amyloid plaques in tissue sections.
  • For Amyloid plaques both intraneuronal and vascular for Amylo-Glo
  • Very bright blue UV excitable stain under physiological conditions
  • Unique excitation/emission profile and mild staining
  • Combination for multiple immunofluorescent labeling studies
  • Storage Up to six months at 4 °C
Amylo-Glo RTD is compatible with fresh, frozen, and formalin-fixed immunohistochemistry or cytochemistry, and particularly good for confocal and multiple labeling because of its high fluorescent intensity and resistance to photo-bleaching.
Moreover because Amylo-Glo fluoresces in the UV channel, double and triple labeling experiments can be performed very easily.
Ethidium bromide is used as a counter stain to visualise cell nuclei and cell bodies of cells while under UV illumination allowing the assessment of amyloid plaques and cell/tissue positioning as well in one step.
Pozor: Protect from light. No preservatives. Use sterile technique when handling and proper laboratory procedures.