Tlačiť

Anti-Protein painting of fourth Rabbit Polyclonal Antibody

Dodávateľ: BIOSENSIS PTY
R-1571-100
BSENR-1571-100EA 356.1 EUR
BSENR-1571-100
Anti-Protein painting of fourth Rabbit Polyclonal Antibody
Protilátky
Protein painting of fourth (Pof) is a probable RNA-binding protein that binds to the fourth chromosome and may bind a RNA that spreads the fourth chromosome (Ref: Swissprot).

Antibody recognizes painting of fourth protein from flies by ELISA. Other applications and other species not yet tested.
Application Information:
ELISA. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Protein painting of fourth
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Drosophila melanogaster Protein painting of fourth (22-36More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR