Tlačiť

Anti-CALB2 Chicken Polyclonal Antibody

Dodávateľ: BIOSENSIS PTY
C-1801-50
BSENC-1801-50EA 356.1 EUR
BSENC-1801-50
Anti-CALB2 Chicken Polyclonal Antibody
Protilátky
Calretinin is a calcium-binding protein which is abundant in auditory neurons. Ref: uniprot.org. Antibody is specific for calretinin and does not recognize closely related proteins parvalbumin and calbindin as determined by Western Blotting.

Western blotting (1:1,000-1:5,000) and Immunohistochemistry (1:1,000-1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CALB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat, Cow
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR