Tlačiť

Anti-Uncoupling Protein 3 Chicken Polyclonal Antibody

Dodávateľ: BIOSENSIS PTY
C-1542-100
BSENC-1542-100EA 366.11 EUR
BSENC-1542-100
Anti-Uncoupling Protein 3 Chicken Polyclonal Antibody
Protilátky
Uncoupling Protein 3 (UCP3) belongs to the mitochondrial carrier family. Located in the mitochondrion inner membrane, UCP3 creates proton leaks across the membrane thus uncoupling oxidative phosphorylation (Ref: SWISSPROT).

This antibody detects UCP3 at approx 35 kDa.
Application Information:
WB. Suggested dilution at 1:1,000 to 1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Uncoupling Protein 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminus of Human Uncoupling Protein 3 (298-312 aa).
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR