Tlačiť
Pipe Marker, Compressed Air
Dodávateľ: BRADY
N004705
BRDYN004705EA 18.55 EUR
BRDYN004705
Pipe Marker, Compressed Air
Značení bezpečnostní
Individual pipe markers on map with perforated arrows, without pictogram.
  • Material: Self-adhesive laminated polyester
  • Colour: White on blue
  • Weatherproof; UV and chemical resistant
Available in multiple sizes and on or without a liner.