Tlačiť
Code Reader™ CR1100 Barcode Scanners
Dodávateľ: BRADY
176692 176693
BRDY176692EA 490.69 EUR
BRDY176692 BRDY176693
Code Reader™ CR1100 Barcode Scanners
Systémy na čárové kódy
Automate your operations with the CR1100. With industrial-grade durability and consumer-grade functionality, you get a seamless experience with every scan.
  • Read more types of barcodes with patented dual-field optics
  • Get extra protection to seal out dust and moisture with an IP54 rating
  • Lightweight and compact with rugged design
  • Is customisable for workflow needs with visual, audible and haptic indicators
  • Features a powerful JavaScript platform for complete device control
  • Includes a stand to protect scanner while not in use
Application: Asset tracking,More Product Information