Tlačiť
Combined lock storage / group lockout boxes
Dodávateľ: BRADY
105716
BRDY105716EA 156.62 EUR
BRDY105716
Combined lock storage / group lockout boxes
Zámky bezpečnostní
These handy boxes feature a lockable storage compartment and a group lockout compartment. Made of heavy duty steel, powder coated for extra rust resistance.
  • Group lockout compartment has clear Lexan window with keyhole slot
  • Storage compartment holds approx. 50 or 125 padlocks
  • 17 or 34 padlocks holes in group lock box lid
Max. shackle diameter: 7,5 mm
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable