Tlačiť
Closing cones, Combi-Stoppers
Dodávateľ: BRAUN
4495101 4495152 4495209
612-0100EA 52.87 EUR
612-0100 BRAU4495152 BRAU4495209
Closing cones, Combi-Stoppers
Zátky Zátky PE/LDPE/HDPE
These PE Combi-Stoppers are used for sealing male or female Luer-Lock and Luer-Slip connections.
  • Unique multifunctional design
  • Sterile closing of pre-filled syringes and all kinds of open IV-accesses in IV-Sets
Available in red, blue and white.