Tlačiť
Infusion valve, Safeflow®
Dodávateľ: BRAUN
409100H
BRAU409100HEA 217.1 EUR
BRAU409100H
Infusion valve, Safeflow®
Infuze
This capless valve allows for a safe and convenient access during infusion therapy.
  • Rapid delivery
  • Compatible with luer-slip and -lock
  • Provides needle-free IV access while eliminating the risk of needle stick injuries
Suitable for every IV procedure.