Tlačiť

Chlorid sodný 0,9%, Ecotainer®

Dodávateľ: B. Braun

Synonymá: Saline, NaCl, Sea salt, Halite

3570130 3570160
BRAU3570130EA 26.84 EUR
BRAU3570130 BRAU3570160
Chlorid sodný 0,9%, Ecotainer®
Chlorid sodný

Sterile and pyrogen-free topical rinse solution.


  • PVC- and latex-free
  • Sterile and non-pyrogenic
  • Handy container with screw cap


Ecotainer® has been designed for intra- and post-operative irrigation.

Vzorec: NaCl
Mw: 58.44
Teplota varu: 1413
Teplota tavenia: 801
Číslo MDL: MFCD00003477
Číslo CAS: 7647-14-5
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR