Tlačiť
Anti-RNF23 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Dodávateľ: BIOSS INC
BS-9238R-A750
BOSSBS-9238R-A750EA 1018.63 EUR
BOSSBS-9238R-A750
Anti-RNF23 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. RNF23 (RING finger protein 23), also known as tripartite motif-containing protein 39 (TRIM39) or testis-abundant finger protein, is a 518 amino acid protein belonging to the TRIM/RBCC family that is known to interact with MOAP1. Ubiquitously expressed and existing as two alternatively spliced isoforms,More Product Information