Tlačiť

Anti-WNT7A Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Dodávateľ: BIOSS INC
BS-6645R-A750
BOSSBS-6645R-A750EA 1018.63 EUR
BOSSBS-6645R-A750
Anti-WNT7A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. Signaling by Wnt-7a allows sexually dimorphic development of the mullerian ducts (By similarity).

Type: Primary
Antigen: WNT7A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 241-349/349
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR